missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Frizzled-1 (NP_003496) VersaClone cDNA
FZD1
Brand: R&D Systems™ RDC0117
Additional Details : Weight : 0.00970kg
Description
The Human Frizzled-1 (NP_003496) VersaClone cDNA clone contains the complete ORF for Frizzled-1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Frizzled-1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hFZD1 | |
10 μg at 0.2 μg/μL | |
1956bp | |
Human |
NP_003496 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans