missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat GDF-11/BMP-11 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

£382.00

Specifications

Gene ID (Entrez) 10220
Molecular Weight (g/mol) 12.6 kDa (monomer)
Name GDF-11/BMP-11
Quantity 10 μg
Source E. coli-derived GDF-11/BMP-11 protein Asn299-Ser407,with an N-terminal Met
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18389707
View Documents
R&D Systems™
1958-GD-010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
£382.00
10µg
Please to purchase this item. Need a web account? Register with us today!
18137890
View Documents
R&D Systems™
1958-GD-010/CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
£382.00
10µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Human/Mouse/Rat GDF-11/BMP-11 Protein is derived from E. coli. The Recombinant Human/Mouse/Rat GDF-11/BMP-11 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

10220
GDF-11/BMP-11
E. coli-derived GDF-11/BMP-11 protein Asn299-Ser407,with an N-terminal Met
Disulfide-linked homodimer
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
12.6 kDa (monomer)
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
BMP11, BMP-11, BMP-11BMP11Bone morphogenetic protein 11, GDF11, GDF-11, growth differentiation factor 11, growth/differentiation factor 11
Recombinant
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.