missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human GPR12 (NP_005279) VersaClone cDNA
G Protein-coupled Receptor 12
Brand: R&D Systems™ RDC0303
Additional Details : Weight : 0.00970kg
Description
The Human GPR12 (NP_005279) VersaClone cDNA clone contains the complete ORF for GPR12, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GPR12 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hGPR12 | |
10 μg at 0.2 μg/μL | |
1018bp | |
Human |
NP_005279 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans