missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human GPR15 (NP_005281) VersaClone cDNA
G Protein-coupled Receptor 15
Brand: R&D Systems™ RDC0064
Additional Details : Weight : 0.00970kg
Description
The Human GPR15 (NP_005281) VersaClone cDNA clone contains the complete ORF for GPR15, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GPR15 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hGPR15 | |
10 μg at 0.2 μg/μL | |
1096bp | |
Human |
NP_005281 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans