missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-2 Protein
Version with mutation found in aldesleukin Available!
£77.80 - £969.00
Specifications
Accession Number | P60568 |
---|---|
For Use With (Application) | Bioactivity |
Gene ID (Entrez) | 3558 |
Molecular Weight (g/mol) | 13 kDa, under reducing conditions. |
Name | IL-2 |
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
18116540
|
R&D Systems™
202-IL-010/CF |
10 μg | Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. |
£78.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18374018
|
R&D Systems™
202-IL-010 |
10 μg | Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. |
£77.80
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18370328
|
R&D Systems™
202-IL-050/CF |
50 μg | Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA. |
£188.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18157790
|
R&D Systems™
202-IL-050 |
50 μg | Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. |
£188.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18624749
|
R&D Systems™
202-IL-500 |
500 μg | Lyophilized from a 0.2 μm filtered solution in acetonitrile and TFA with BSA as a carrier protein. |
£969.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Measured in a cell proliferation assay using CTLL-2 mouse cytotoxic T cells. Gearing, A.J.H. and C.B. Bird (1987) in Lymphokines and Interferons, A Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 295. The ED50 for this effect is 0.05-0.25 ng/mL. The specific activity of Recombinant Human IL-2 is approximately 2.1 x 104 IU/μg, which is calibrated against recombinant human IL-2 WHO International Standard (NIBSC code: 86/500).Specifications
P60568 | |
3558 | |
IL-2 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
IL2 | |
Recombinant | |
IL-2 |
Bioactivity | |
13 kDa, under reducing conditions. | |
E. coli-derived human IL-2 protein Ala21-Thr153, with an N-terminal Met | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |