Learn More
R&D Systems™ Human IL-3R alpha/CD123 (NP_002174) VersaClone cDNA
Interleukin 3 Receptor alpha
Brand: R&D Systems™ RDC0896
This item is not returnable.
View return policy
Description
The Human IL-3 R alpha/CD123 (NP_002174) VersaClone cDNA clone contains the complete ORF for IL-3R alpha/CD123, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The IL-3R alpha/CD123 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hIL3RA | |
10 μg at 0.2 μg/μL | |
1150bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
NP_002174 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 R alpha, IL-3 receptor subunit alpha, IL3R, IL3R alpha, IL-3R subunit alpha, IL3RA, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 | |
Human |
Product Content Correction
Product Title
For Reasearch Use Only; Not for Use in Humans
Spot an opportunity for improvement?Share a Content Correction