Learn More
R&D Systems™ Human MCT8/SLC16A2 (NP_006508) VersaClone cDNA
Monocarboxylic Acid Transporter 8 (Solute Carrier Family 16 Member 1)
Brand:  R&D Systems™ RDC0892
Product Code. 18732559
  • £281.00 / 10µg
Explore more special offers
Description
Description
The Human MCT8/SLC16A2 (NP_006508) VersaClone cDNA clone contains the complete ORF for MCT8/SLC16A2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The MCT8/SLC16A2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
hSLC16A2
10 μg at 0.2 μg/μL
1632bp
Human
NP_006508
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&D Systems™ Human MCT8/SLC16A2 (NP_006508) VersaClone cDNA > 10ug
For Reasearch Use Only; Not for Use in Humans