Learn More
R&D Systems™ Human Mesenchymal Stem Cell Functional Identification Kit
To verify multipotency of human mesenchymal stem cells by in vitro functional differentiation
Brand:  R&D Systems™ SC006
Product Code. 18712144
  • £416.00 / 1 set
Explore more special offers
Includes: Adipogenic Supplement, Osteogenic Supplement, Chondrogenic Supplement, ITS Supplement, Goat anti-mouse FABP-4, Mouse anti-human Osteocalcin, Goat anti-human Aggrecan
Description
Description
Can be used with the Human MSC Verification Flow Kit (Mfr. No. FMC020) to define MSCs according to the ISCT recommendations
Reliably induces MSC trilineage differentiation using defined supplements
Includes premium quality antibodies to confirm differen
Specifications
Specifications
1 Kit
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.