missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Proprotein Convertase 7/PCSK7 Protein, CF

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 2984-SE-010

Product Code. 18347407

  • £385.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured by its ability to cleave the fluorogenic peptide substrate pERTKR-AMC (Catalog # ES013 ). The specific activity is >125 pmol/min/μg, as measured under the described conditions.
Specifications

Specifications

Q16549
Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl2 and Glycerol.
M.W. (Observed): 79 kDa, reducing conditions; M.W. (theoretical): 70 kDa (pro form) & 59 kDa (mature form)
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
PCSK7
Recombinant
Proprotein Convertase 7/PCSK7
Enzyme Activity
9159
Proprotein Convertase 7/PCSK7
Mouse myeloma cell line, NS0-derived human Proprotein Convertase 7/PCSK7 protein Leu38-Thr667, with a 10-His tag between Arg141 and Ser142
<1.0 EU / 1 μg of the protein by the LAL method.
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.