missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Trypsin 2/PRSS2 Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 3586-SE-010
234.12 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (Catalog # ES002 ). The specific activity is >8,000 pmol/min/μg, as measured under the described conditions.Specifications
P07478 | |
Supplied as a 0.2 μm filtered solution in HCl and NaCl. | |
M.W. (Observed): 34 kDa, reducing conditions; M.W. (theoretical): 26 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain |
Enzyme Activity | |
5645 | |
Trypsin 2/PRSS2 | |
Mouse myeloma cell line, NS0-derived human Trypsin 2/PRSS2 protein Ala16-Ser247, with a C-terminal 10-His tag | |
Pro form | |
PRSS2 | |
Recombinant | |
Trypsin 2/PRSS2 |