Learn More
R&D Systems™ Human TSPAN32/TSSC6 (NP_620591) VersaClone cDNA
Tetraspanin 32
Brand: R&D Systems™ RDC0890
This item is not returnable.
View return policy
Description
The Human TSPAN32/TSSC6 (NP_620591) VersaClone cDNA clone contains the complete ORF for TSPAN32/TSSC6, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TSPAN32/TSSC6 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hTSPAN32 | |
10 μg at 0.2 μg/μL | |
976bp | |
Human |
NP_620591 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans