missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human VEGFR1/Flt-1 (NP_002010) VersaClone cDNA
Vascular Endothelial Growth Factor Receptor 1
Brand: R&D Systems™ RDC0466
Additional Details : Weight : 0.00970kg
Description
The Human VEGFR1/Flt-1 (NP_002010) VersaClone cDNA clone contains the complete ORF for VEGFR1/Flt-1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The VEGFR1/Flt-1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
hVEGFR1 | |
10 μg at 0.2 μg/μL | |
4030bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
NP_002010 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
EC 2.7.10, EC 2.7.10.1, FLT, FLT1, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGF R1, VEGFR1, VEGFR-1 | |
Human |
For Reasearch Use Only; Not for Use in Humans