missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mca-PLAQAV-Dpa-RSSSR-NH2 Fluorogenic Peptide Substrate

Applications: Bioactivity

Brand:  R&D Systems™ ES003

222.89 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18136380

  • £292.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2, 4-dinitrophenyl group
  • It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence
  • Also an excellent substrate for ADAM8

Specifications

Specifications

Liquid
Supplied as a stock solution in dimethyl sulfoxide (DMSO) at a concentration of 8.0mg/mL or 4 mM.
Source: Synthetic
Bioactivity
Predicted MW: 1638 Da
Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers