missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mca-PLGL-Dpa-AR-NH2 Fluorogenic MMP Substrate

Applications: Bioactivity, Binding Activity

Brand:  R&D Systems™ ES001

Product Code. 18116670

  • £292.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Substrate for MMP-1, MMP-2, MMP-7, MMP-8, MMP-9, MMP-12, MMP-13, MMP-14, MMP-15, MMP-16, Cathepsin D, and Cathepsin E
  • Contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2, 4-dinitrophenyl group
  • It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence
  • Also an excellent substrate for active Cathepsins D and E
  • Purity 98±1% based on HPLC

Specifications

Specifications

Liquid
Supplied as a stock solution in dimethyl sulfoxide (DMSO) at a concentration of 7.99mg/mL or 6.07 mM.
Source: Synthetic
Bioactivity, Binding Activity
Predicted MW: 1093 Da
Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.