missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mca-RPPGFSAFK(Dnp)-OH Fluorogenic Peptide Substrate

Applications: Bioactivity, Binding Activity

Brand:  R&D Systems™ ES005

Product Code. 18345577

  • £292.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2, 4-dinitrophenyl group
  • It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence
  • Purity >95% by HPLC
  • The cleavage site by ECE-1 is the peptide bond between Ala and Phe

Specifications

Specifications

Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Bioactivity, Binding Activity
Predicted MW: 1388 Da
Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Liquid
Stock solution at 4mM or 6.26mg/mL (0.16 mL) dimethyl sulfoxide (DMSO).
Source: Synthetic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.