missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mca-YVADAPK(Dnp)-OH Fluorogenic Peptide Substrate

Applications: Bioactivity

Brand:  R&D Systems™ ES007

222.89 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18339707

  • £292.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Contains a highly fluorescent 7-methoxycoumarin group that is efficiently quenched by resonance energy transfer to the 2, 4-dinitrophenyl group
  • It can be used to measure the activities of peptidases that are capable of cleaving an amide bond between the fluorescent group and the quencher group, causing an increase in fluorescence
  • Purity >95% by HPLC
  • The cleavage sites by ICE and ACE-2 are the peptide bonds between Ala and Asp and between Pro and Lys, respectively

Specifications

Specifications

Liquid
Supplied as a stock solution in dimethyl sulfoxide (DMSO) at a concentration of 6.19mg/mL or 4.0 mM.
Purity: >95%, by HPLC, Source: Synthetic
Bioactivity
Predicted MW: 1145.2 Da
Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers