Learn More
R&D Systems™ Mouse FATP2 (NP_036108) VersaClone cDNA
Fatty Acid Transport Protein 2
Brand:  R&D Systems™ RDC0909
Product Code. 18718129
  • £648.00 / 10µg
Explore more special offers
Description
Description
The Mouse FATP2 (NP_036108) VersaClone cDNA clone contains the complete ORF for FATP2/SLC27A2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The FATP2/SLC27A2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
mSlc27a2
10 μg at 0.2 μg/μL
1875bp
Mouse
NP_036108
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Reasearch Use Only; Not for Use in Humans