Learn More
R&D Systems™ Mouse Mesenchymal Stem Cell Functional Identification Kit
To verify multipotency of mouse mesenchymal stem cells by in vitro functional differentiation
Brand:  R&D Systems™ SC010
Product Code. 18761814
  • £551.00 / 1 set
Explore more special offers
Includes: Adipogenic Supplement, Mouse/Rat Osteogenic Supplement, Chondrogenic Supplement, ITS Supplement, Goat anti-mouse FABP4, Sheep Anti-mouse Collagen II, Goat Anti-mouse Osteopontin
Description
Description
Reduces experimental variation
Reliably induces MSC trilineage differentiation using defined supplements
A panel of antibodies, consisting of Goat Anti-mouse FABP4, Sheep Anti-mouse Collagen II, and Goat Anti-mouse Osteopontin, are included to define th
Specifications
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
1 Kit