missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse MS4A4C (NP_083775) VersaClone cDNA
Membrane-spanning 4-domains, Subfamily A, Member 4C
Brand: R&D Systems™ RDC0235
Additional Details : Weight : 0.00970kg
Description
The Mouse MS4A4C (NP_083775) VersaClone cDNA clone contains the complete ORF for MS4A4C, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The MS4A4C insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
mMS4A4C | |
10 μg at 0.2 μg/μL | |
694bp | |
Mouse |
NP_083775 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans