missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse SECTM1B (NP_081183) VersaClone cDNA
Secreted and Transmembrane 1B
Brand: R&D Systems™ RDC0740
Additional Details : Weight : 0.00970kg
Description
The Mouse SECTM1B (NP_081183) VersaClone cDNA clone contains the complete ORF for SECTM1B, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SECTM1B insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
mSECTM1B | |
10 μg at 0.2 μg/μL | |
652bp | |
Mouse |
NP_081183 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans