Learn More
R&D Systems™ MycoProbe Mycoplasma Detection Kit
A complete kit to detect common antibiotic-resistant cell culture contaminants
Brand: R&D Systems™ CUL001B
386.17 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Mycoplasma contamination affects up to 80% of continuous cell cultures. If undetected, mycoplasma contamination can have significant effects on the quality and reliability of cell culture preparations.
Reduces experimental variation
Accurate and highly sensitive
Simple multiwell-based colorimetric assay
Only takes 4.5 hours
Specifications
Store the unopened product at 2 - 8° C. Do not use past expiration date. |