Learn More
R&D Systems™ N21-MAX Media Supplement (50X)
For long-term culture of neurons under fully defined, serum-free conditions
Brand: R&D Systems™ AR008
74.30 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Description
Serum-free, defined media are routinely used as an alternative to standard serum-containing media in order to reduce unwanted experimental variability. Uncontrolled variables commonly associated with serum-supplemented media, such as indeterminate levels of vitamins, hormones, and growth factors are eliminated, and the potential for contamination by infectious agents is reduced when cells are cultured under defined conditions. In addition, serum-free media offers researchers the ability to specifically design the culture media for a particular experimental question.
- Is fully defined to reduce unwanted experimental variation
- Contains high quality factors to support reproducible and long-term neuron culture
- Does not require an astrocyte feeder layer
- Has been tested to support the viability and growth of E18 rat hippocampal neurons
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 mL |