Learn More
R&D Systems™ pERTKR-AMC Fluorogenic Peptide Substrate
Substrate for Furin, Proprotein Convertase 1 and 7, and wnvNS3
Brand: R&D Systems™ ES013
196.97 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
- Hydrolysis of Arg-AMC amide bond releases AMC, a highly fluorescent group
- A fluorescence plate reader with excitation at 380nm and emission at 460nm is recommended for the measurement of the enzymatic activity
- Purity >96%
- Excellent substrate for furin-like proprotein convertases
Specifications
L-PyroGlu-Arg-Thr-Lys-Arg-AMC | |
Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquoted and stored -20° C to -70° C in a manual defrost freezer for six months. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light. |