Learn More
R&D Systems™ Recombinant Human MMP-1 Western Blot Standard Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Western Blot
Brand:  R&D Systems™ WBC024
125.26 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18327757
  • £165.00 / 100µL
Explore more special offers
Description
Description
The Recombinant Human MMP-1 Western Blot Standard Protein has been validated for the following applications: Western Blot.
Specifications
Specifications
Western Blot
MMP-1 Western Blot Standard
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Unconjugated
MMP-1
4312
1 Vial
MMP1
Recombinant
Product Suggestions