missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human MMP-10 Western Blot Standard Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Western Blot
Brand: R&D Systems™ WBC026
This item is not returnable.
View return policy
Description
The Recombinant Human MMP-10 Western Blot Standard Protein has been validated for the following applications: Western Blot.Specifications
Western Blot | |
MMP-10 Western Blot Standard | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
Unconjugated | |
MMP-10 |
4319 | |
1 Vial | |
MMP10 | |
Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction