missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human MMP-9 Western Blot Standard Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Western Blot

Brand:  R&D Systems™ WBC018

Product Code. 18116250

  • £170.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human MMP-9 Western Blot Standard Protein is derived from CHO. The Recombinant Human MMP-9 Western Blot Standard Protein has been validated for the following applications: Western Blot.
Specifications

Specifications

Western Blot
4318
MMP-9 Western Blot Standard
Chinese Hamster Ovary cell line, CHO-derived human MMP-9 protein
MMP9
Recombinant
MMP-9
No With Carrier Product Bottling Info
M.W. (Observed): 92 kDa
1 Vial
Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Unconjugated
Resuspend the lyophilized powder in 250 uL SDS-PAGE sample buffer.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.