Learn More
R&D Systems™ Resazurin
Simple and quantitative method for measuring cell proliferation, viability and cytotoxicity
Brand:  R&D Systems™ AR002
Product Code. 18411350
  • £169.00 / 100mL
Explore more special offers
Description
Description
  • Nontoxic, water soluble, redox-sensitive dye that changes from its blue/non-fluorescent state to pink/highly-fluorescent upon reduction to resorufin by viable cells
  • Fluorescence can be read using 544nm excitation and 590nm emission wavelengths, or absorbance can be read using a spectrophotometer set at 570nm
  • Resazurin may be added at any time during the culture period
  • For measurement of cell proliferation, it is best to add Resazurin during log phase growth
Specifications
Specifications
100 mL
Store the unopened product at 2 - 8° C. Protect from light. Do not use past expiration date.