Learn More
R&D Systems™ StemXVivo Mesenchymal Stem Cell Expansion Media
A complete, fetal bovine serum-containing media formulated and optimized for the maintenance and expansion of purified human, mouse and rat MSCs
Brand: R&D Systems™ CCM004
Additional Details : Weight : 0.09700kg
Description
- Supports reproducible MSC growth in vitro with a formula to reduce variation
- Ready-to-use formula can be supplemented with user-defined cytokines and growth factors, although these are not needed for culture
- Has been developed and optimized for use with human, mouse, and rat MSCs
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
Stem Cell Expansion Media | |
250 mL |