missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Tyrosine Phosphatase Substrate I, DADEY(PO3)LIPQQG

Substrate for PTP1B, TC-PTP, SHP-2, and Tyrosine Phosphatase

Brand:  R&D Systems™ ES006

222.89 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18379037

  • £292.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Enzymatic cleavage of this substrate liberates inorganic phosphate, which can be measured in dye binding assays, such as the malachite green method
  • Synthetic phosphopeptide corresponding to amino acids 988 - 998 of the mature EGF receptor
  • Derived from an autophosphorylation site at Y992 of EGFR
  • Purity 98% based on HPLC analysis
  • Dissolves in water

Specifications

Specifications

Lyophilized
Predicted MW: 1328.32 Da
Samples are stable for up to twelve months from date of receipt at -20° C to -70° C. The substrate can be aliquotted and stored -20° C to -70° C in a manual defrost freezer for one month. Avoid repeated freeze-thaw cycles. Protect from exposure to direct light.
Lyophilized from a stock solution in distilled water.
Purity: >95%, by HPLC, Source: Synthetic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers