missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Z-FR-AMC Fluorogenic Peptide Substrate
Substrate for Papain
Brand: R&D Systems™ ES009
This item is not returnable.
View return policy
Description
- Hydrolysis of Arg-AMC amide bond releases 7-amino-4-methyl coumarin (AMC), a highly fluorescent group
- A fluorescence plate reader with excitation at 380nm and emission at 460nm is recommended for the measurement of the enzymatic activity
- Often used in characterization of cysteine protease inhibitors
- Purity >99% by thin-layer chromatography
Specifications
Liquid | |
Supplied as a stock solution in dimethyl sulfoxide (DMSO) with a concentration of 100 mg/mL by mass or 157.4 mM with consideration of the peptide content. | |
Z-Phe-Arg-AMC |
Bioactivity | |
Theoretical Molecular weight: 612.6 Da | |
No Stability Data Storage is set to N/A for ProdCode |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction