missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Z-LR-AMC Fluorogenic Peptide Substrate

Applications: Bioactivity

Brand:  R&D Systems™ ES008

180.56 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18108300

  • £237.00 / 10mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The peptide substrate contains a highly fluorescent 7-amino-4-methyl coumarin (AMC) group that is efficiently quenched by the amide bond formed between its amino group and the carboxyl group of the Arg residue
  • Purity 99%±1% by HPLC

Specifications

Specifications

Liquid
Supplied as a stock solution in dimethyl sulfoxide (DMSO)
PredictedMoleMass: 578 Da
Bioactivity
Source: Synthetic
Z-Leu-Arg-AMC. Z: N-carbobenzyloxy; 7-Amino-4-methylcoumarin.
Z: N-carbobenzyloxy; 7-Amino-4-methylcoumarin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers