Learn More
(R)-(-)-Mandelic Acid 99%, Thermo Scientific Chemicals
£15.50 - £72.40
Chemical Identifiers
CAS | 611-71-2 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.15 |
MDL Number | MFCD00064251 |
InChI Key | IWYDHOAUDWTVEP-SSDOTTSWSA-N |
Synonym | r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, --mandelic acid, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid |
PubChem CID | 11914 |
ChEBI | CHEBI:17656 |
IUPAC Name | (2R)-2-hydroxy-2-phenylacetic acid |
SMILES | O[C@@H](C(O)=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10417673
|
Thermo Scientific Acros
125320050 |
5 g | Glass bottle |
£15.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10001290
|
Thermo Scientific Acros
125320250 |
25 g | Plastic bottle |
£26.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10376860
|
Thermo Scientific Acros
125321000 |
100 g | Plastic bottle |
£72.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
611-71-2 | |
152.15 | |
IWYDHOAUDWTVEP-SSDOTTSWSA-N | |
11914 | |
(2R)-2-hydroxy-2-phenylacetic acid |
C8H8O3 | |
MFCD00064251 | |
r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, --mandelic acid, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid | |
CHEBI:17656 | |
O[C@@H](C(O)=O)C1=CC=CC=C1 |
Specifications
611-71-2 | |
98.5 | |
Beige-Yellow to White | |
C8H8O3 | |
MFCD00064251 | |
IWYDHOAUDWTVEP-SSDOTTSWSA-N | |
(2R)-2-hydroxy-2-phenylacetic acid | |
11914 | |
152.15 | |
Authentic | |
(R)-(-)-Mandelic acid, 99% |
130°C to 135°C | |
100.0 | |
99% | |
C6H5CH(OH)CO2H | |
r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, --mandelic acid, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid | |
O[C@@H](C(O)=O)C1=CC=CC=C1 | |
152.15 | |
CHEBI:17656 | |
≥98.5% | |
Crystalline Powder, Crystals or Flakes |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
LIGHT SENSITIVE
missing translation for 'einecsNumber' : 210-276-6
missing translation for 'tsca' : TSCA
- (R)-(-)-Mandelic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only