missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-N-FMOC-(2-Furyl)alanine, 95%, 98% ee, Thermo Scientific™
£70.40 - £129.09
Chemical Identifiers
CAS | 220497-85-8 |
---|---|
Molecular Formula | C22H19NO5 |
Molecular Weight (g/mol) | 377.40 |
MDL Number | MFCD01311747 |
InChI Key | AJXDCHXGNUFBRC-UHFFFAOYSA-N |
Synonym | fmoc-d-2-furylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-d-ala-oh, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-3-2-furyl-d-alanine, n-fmoc-3-2-furyl-d-alanine, fmoc-beta-2-furyl-d-ala-oh hplc |
PubChem CID | 16211163 |
IUPAC Name | (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid |
SMILES | OC(=O)C(CC1=CC=CO1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10743603
|
Thermo Scientific Acros
362495000 |
500 mg | Glass bottle |
£70.40
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10282472
|
Thermo Scientific Acros
362490010 |
1 g | Glass bottle | ||||||
Description
Chemical Identifiers
220497-85-8 | |
377.40 | |
AJXDCHXGNUFBRC-UHFFFAOYSA-N | |
16211163 | |
OC(=O)C(CC1=CC=CO1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
C22H19NO5 | |
MFCD01311747 | |
fmoc-d-2-furylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-d-ala-oh, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-3-2-furyl-d-alanine, n-fmoc-3-2-furyl-d-alanine, fmoc-beta-2-furyl-d-ala-oh hplc | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid |
Specifications
220497-85-8 | |
100.0 | |
97.5% min. (HPLC) | |
95% | |
MFCD01311747 | |
AJXDCHXGNUFBRC-UHFFFAOYSA-N | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid | |
16211163 | |
≥94% | |
(R)-N-FMOC-(2-Furyl)alanine, 95%, (98% e.e.) |
94.0 | |
White to Yellow | |
Authentic | |
C22H19NO5 | |
fmoc-d-2-furylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-d-ala-oh, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-3-2-furyl-d-alanine, n-fmoc-3-2-furyl-d-alanine, fmoc-beta-2-furyl-d-ala-oh hplc | |
OC(=O)C(CC1=CC=CO1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
377.40 | |
377.39 | |
Powder |
Safety and Handling
missing translation for 'tsca' : TSCA
RUO – Research Use Only