missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-Propranolol hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
100 mg
Description
Less active enantiomer of the β-adrenoceptor antagonist propranolol .
Racemate, S-enantiomer and Glycol Metabolite also available.
Specifications
Specifications
CAS | 13071-11-9 |
Molecular Formula | C16H22ClNO2 |
Quantity | 100 mg |
Synonym | r-+-propranolol hydrochloride, dexpropranolol hydrochloride, dexpropranolol hcl, unii-ab9db0nx46, ab9db0nx46, dexpropranolol hydrochloride usan, r-1-isopropylamino-3-1-naphthyloxy-2-propanol hydrochloride, 2r-1-naphthalen-1-yloxy-3-propan-2-ylamino propan-2-ol hydrochloride, +-anapriline |
InChI Key | ZMRUPTIKESYGQW-PFEQFJNWSA-N |
SMILES | CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl |
IUPAC Name | (2R)-1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride |
Molecular Weight (g/mol) | 295.807 |
PubChem CID | 66366 |
ChEBI | CHEBI:51161 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'