Learn More
R&D Systems™ Quantikine Immunoassay Control Set 665 for Human Uteroglobin
For use with catalog number DUGB00
Brand: R&D Systems™ QC181
This item is not returnable.
View return policy
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date. | |
7356 | |
1 Each |
Uteroglobin/SCGB1A1 | |
Control | |
Human |