missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 725 for Human PDGF-DD

Product Code. 30001929
Click to view available options
Quantity:
1 Each
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30001929

Brand: R&D Systems™ QC140

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with catalog number DDD00

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes PDGF-DD
Gene ID (Entrez) 80310
Intended Use Control
Quantity 1 Each
Source Human
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.