missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ (R,R)-Chloramphenicol, 98% Product Code.: 11445786

Thermo Scientific™ (R,R)-Chloramphenicol, 98%

Product Code. 11445786
25g
Click to view available options
Quantity:
25g
100g
This item is not returnable. View return policy

Product Code. 11445786

Brand: Thermo Scientific™ B20841.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(R,R)-Chloramphenicol, CAS # 56-75-7, also known as chloromycetin, is a compound with broad-spectrum antibiotic and bacteriostatic activities.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

  • (R,R)-chloramphenicol is a compound with antibiotic and bacteriostatic activity and is derived from Streptomyces venequelae
  • It that can block the translation on the 50S subunit at the peptidyltransferase step. The binding affects the peptidyl transferase activity, preventing the transfer of amino acids to the growing peptide chains and inhibiting peptide bond formation. The resulting inhibition of the bacterial protein synthesis impedes bacterial cell proliferation

Application

  • (R,R)-chloramphenicol has been used for several molecular biology approaches, mainly for bacterial selection. It can be used to select transformed cells containing chloramphenicol resistance genes
  • This compound can also promote mitochondrial and chloroplast protein synthesis and the ribosomal formation of (p)ppGpp, depressing the rRNA transcription process
  • In vitro results demonstrate that (R,R)-chloramphenicol shows activity against several vancomycin-resistant E. faecium strains
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-75-7
Molecular Formula C11H12Cl2N2O5
Molecular Weight (g/mol) 323.126
MDL Number MFCD00078159
InChI Key WIIZWVCIJKGZOK-RKDXNWHRSA-N
PubChem CID 5959
ChEBI CHEBI:17698
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
SMILES C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]

Specifications

Quantity 25g
Assay Percent Range 0.99
Beilstein 2225532
Merck Index 14,2077
Solubility Information Soluble in alcohol,propylene,glycol,acetone and ethyl acetate. Slightly soluble in water.
Formula Weight 323.14
Percent Purity ≥99%
Chemical Name or Material Chloramphenicol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX21K020 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberB20841.14,B20841.22
Lot NumberR20K026 Certificate TypeCertificate of Analysis Date24/05/2024 Catalog NumberB20841.14,B20841.22
Lot NumberM05K042 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog NumberB20841.14,B20841.22
Lot Number10214539 Certificate TypeCertificate of Analysis Date03/07/2023 Catalog NumberB20841.14,B20841,B20841.22
Lot NumberY07I031 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog NumberB20841.14,B20841.22
compliance-icons
Product Identifier
  • (R,R)-Chloramphenicol
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Chloramphenicol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ (R,R)-Chloramphenicol, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.