missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-S-Methyl-S-phenylsulfoximine, 97%, Thermo Scientific Chemicals

Product Code. 11332586
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 60933-65-5
Molecular Formula C7H9NOS
Molecular Weight (g/mol) 155.215
MDL Number MFCD00151460
InChI Key YFYIDTVGWCYSEO-JTQLQIEISA-N
Synonym s-+-s-methyl-s-phenylsulfoximine, s-+-s-methyl-s-phenylsulfoximine ee, s-imino methyl phenyl-??-sulfanone, r---s-methyl-s-phenylsulfoximine ee, imino-methyl-oxo-phenyl-$l^ 6, s-+-s-methyl-s-phenylsulphoximine, sulfoximine,s-methyl-s-phenyl-, s s
PubChem CID 25036288
IUPAC Name imino-methyl-oxo-phenyl-$l^{6}-sulfane
SMILES CS(=N)(=O)C1=CC=CC=C1

Spécification

Melting Point 29°C to 31°C
Quantity 1 g
Beilstein 5245635
Sensitivity Hygroscopic
Optical Rotation −36° (c=1 in Acetone)
Formula Weight 155.22
Percent Purity 97%
Chemical Name or Material (R)-(-)-S-Methyl-S-phenylsulfoximine

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis