Learn More
Brady™ R10000 Series Printer Ribbon
Switch out your ribbon in seconds with these easy-to-load ribbon cartridges that slide into place in the always-correct direction.
Brand:  Brady™ 151001
 View more versions of this product
Product Code. 17595126
  • £73.70 / Each
Estimated Shipment: 10-12-2024
to see stock.
Découvrez plus d'offres spéciales
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
  • Make it last – R10000 series ribbons are based on a resin formulation that produces long-lasting printed text that can withstand years in industrial areas and outdoor conditions
  • Make it fast – self-contained ribbon cartridges are easy to replace eliminating the hassle of printer calibration and ribbon threading
  • R10000 series ribbons are ideal for use with long-lasting indoor / outdoor materials such as Brady B-595
Spécification
Spécification
White
Resin
75 ft.
75 ft. x 2 in.
BMP61 Labeling System, BradyPrinter M611
Thermal Transfer
2 in.
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats