Learn More
Gilson™ Minipuls™ 3 R8 Medium-Flow Pump Head
For use with Peristaltic pump
Brand: Gilson™ F117608
1756.03 GBP valid until 2024-11-30
Use promo code "21681" to get your promotional price.
This item is not returnable.
View return policy
Description
- Standard model
- With 10 stainless steel rollers per head
- Eight channels
Specifications
0.3μL to 30 mL/min. | |
8 | |
Stainless Steel |
R8 medium flow pump head | |
10 | |
Peristaltic Pump Head |