Learn More
Epredia™ RA Lamb BlockStore Storage Options for the LabStack Storage System
Store lab supplies that don't typically fit in standard-size compartments with this versatile, expandable system.
£651.00
Product Code | Brand | Product Type | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Color | Price | Quantity & Availability | ||||
12181830
|
Epredia™
E43/22GREY |
BlockStore 8-Drawer Cabinet | Gray |
£651.00
Each |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Compatibility:
- Compatible with all standard LabStack cabinets and options
- Accepts embedding rings, processing cassettes, and unmounted paraffin blocks
Includes:
- 8-Drawers (arranged in two rows of four)
- Enough dividers to produce seven rows, 1.2 in. (30 mm) wide, in each drawer
Specifications
Gray |