missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528032
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (98%), Rat (97%), Xenopus laevis (88%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Protein transport. Probably involved in vesicular traffic. Target protein may play a role in secretory granule function and exocytosis in pancreatic acinar cells.
Specifications
| RAB3B | |
| Polyclonal | |
| Unconjugated | |
| Rab3b | |
| 2610528C18Rik; brain antigen RAB3B; Rab3b; RAB3B, member RAS oncogene family; ras-related protein Rab-3B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5865 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P20337 | |
| Rab3b | |
| Recombinant protein encompassing a sequence within the center region of human RAB3B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction