missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB5A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11947-1-AP-150UL
This item is not returnable.
View return policy
Description
RAB5 are among the small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different sets of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. RAB5A is required for the fusion of plasma membranes and early endosomes. Contributes to the regulation of filopodia extension. Required for the exosomal release of SDCBP, CD63, PDCD6IP and syndecan. Regulates maturation of apoptotic cell-containing phagosomes, probably downstream of DYN2 and PIK3C3.Specifications
RAB5A | |
Polyclonal | |
Unconjugated | |
RAB5A | |
RAB5, RAB5A, Ras related protein Rab 5A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
271457, 5868, 64633 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
M0RC99, P20339, Q9CQD1 | |
RAB5A | |
RAB5A Fusion Protein Ag2549 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |