Learn More
RAB7A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55469-1-AP-20UL
This item is not returnable.
View return policy
Description
The RAB7A antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human RAB7A. This antibody recognizes human, mouse, rat antigen. The RAB7A antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
RAB7A | |
Polyclonal | |
Unconjugated | |
20 μL | |
7879 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry | |
380 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |