missing translation for 'onlineSavingsMsg'
Learn More

BNIP3 Rabbit anti-Chicken, Clawed frog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of BNIP3.

Brand:  Abnova PAB0264.50uL

Product Code. 16088666

  • £423.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This gene is a member of the BCL2/adenovirus E1B 19kDa-interacting protein (BNIP) family. It interacts with the E1B 19kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19kDa-like sequences of BCL2, also an apoptotic protector. This gene contains a BH3 domain and a transmembrane domain, which have been associated with pro-apoptotic function. The dimeric mitochondrial protein encoded by this gene is known to induce apoptosis, even in the presence of BCL2. [provided by RefSeq

Sequence: SRPENIPPKEFLFKHPKR
Specifications

Specifications

BNIP3
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of BNIP3.
In serum (0.05% sodium azide)
NIP3
Rabbit
50 μL
Primary
Chicken, Frog, Human, Mouse, Rat
Liquid
Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000-1:5000) Immunohistochemistry (Frozen sections) Immunoprecipitation (1:50-1:200) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the
BNIP3
BNIP3
A synthetic peptide corresponding to amino acids 122-139 of human BNIP3.
RUO
664
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only