Learn More
EMR1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of EMR1.
Brand: Abnova PAB26229.50ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a protein that has a domain resembling seven transmembrane G protein-coupled hormone receptors (7TM receptors) at its C-terminus. The N-terminus of the encoded protein has six EGF-like modules, separated from the transmembrane segments by a serine/threonine-rich domain, a feature reminiscent of mucin-like, single-span, integral membrane glycoproteins with adhesive properties. [provided by RefSeq
Specifications
EMR1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of EMR1. | |
In PBS (0.09% sodium azide) | |
Q14246 | |
EMR1 | |
A synthetic peptide corresponding to 16 amino acid at N-terminus of human EMR1. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10-20 ug/mL) The optimal working dilution should be determined by the end user. | |
EMR1 | |
TM7LN3 | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
2015 | |
Human | |
Liquid |
For Research Use Only