missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat IgG (H+L) Secondary Antibody, HRP, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen 611620
Code : 50
Additional Details : Weight : 0.50000kg
Description
Goat IgG (H+L) Polyclonal antibody specifically detects Goat IgG (H+L) in Goat samples. It is validated for ELISA, Western Blot.Specifications
Goat IgG (H+L) | |
Polyclonal | |
HRP | |
Rabbit | |
Affinity chromatography | |
RUO | |
Goat | |
Liquid |
ELISA, Western Blot | |
1.5 mg/mL | |
PBS with 40% glycerol, 4mg/mL BSA and 0.19% Kathon™ CG; pH 7.4 | |
Gamma Immunoglobin | |
3 mg | |
Secondary | |
4° C | |
IgG |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Rabbit anti-Goat IgG (H+L) Secondary Antibody, HRP
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 5-Chloro-2-methyl-3(2H)-isothiazolone