missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Horse serum proteins, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB9272.2mL
Additional Details : Weight : 0.00010kg
Specifications
Rabbit anti-Horse serum proteins | |
Polyclonal | |
Rabbit anti-horse IgG recognizes horse serum proteins. This secondary antibody was purified using delipidation and defibrination. | |
Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
Horse serum proteins | |
2 mL | |
Equine | |
Lyophilized |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
This product was prepared from polyspecific antiserum by a delipidationand defibrination. Assay by immunoelectrophoresis resulted in a multipleprecipitin arcs against horse serum. | |
Secondary | |
Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only