missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Human Ig (Light chain) Recombinant Secondary Antibody, Biotin, Invitrogen™

Rabbit Recombinant Monoclonal Secondary Antibody

Brand:  Invitrogen SA510224

Product Code. 15869893

  • £305.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This antibody reacts to both kappa and lambda light chains of human immunoglobulins. No cross reactivity with mouse, rat, or goat immunoglobulin light chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Human Ig (Light chain)
Recombinant Monoclonal
1.0 mg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH 7.2-7.4
Peptide corresponding to Human IgG.
50 μg
Secondary
-20° C, Avoid Freeze/Thaw Cycles
IgG
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry
RM129
Biotin
Rabbit
Protein A
RUO
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only