missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 800, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen SA510116
This item is not returnable.
View return policy
Description
This antibody is cross-adsorbed and exhibits minimum reactivity to mouse and rat.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Human IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
DyLight 800 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 | |
Human IgG-heavy and light chain | |
500 μg | |
Secondary | |
4° C | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction